Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • COPII Antibodies

          Invitrogen

          COPII Polyclonal Antibody

          10 Published Figures
          19 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite COPII Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (10)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          COPII Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          COPII Antibody (PA1-069A) in ICC/IF

          Immunofluorescent analysis of COP II (green) showing staining in the cytoplasm of NIH-3T3 cells (right) compared to a negative control without primary antibody (left). Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a COP II polyclonal antibody (Product # PA1-069A) in 3% BSA-PBS at a dilution of 1:100 and incubated overnight at 4 °C in a humidified chamber. Cells were washed with PBST and incubated with a DyLight-co... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunocytochemistry (ICC/IF)
          COPII Antibody in Immunohistochemistry (IHC)
          COPII Antibody in Western Blot (WB)
          COPII Antibody in Western Blot (WB)
          COPII Polyclonal Antibody

          Product Details

          PA1-069A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:200
          View 10 publications 10 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Fruit fly, Hamster, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2301581

          Product Specific Information

          PA1-069A detects coatomer-protein II (COPII) from rat, mouse and human cells.

          PA1-069A has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ~85 kDa protein representing COPII from PC-12 cell extract.

          The PA1-069A immunogen is a synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII. This sequence is completely conserved between human, mouse, and rat. PA1-069A immunizing peptide (Cat. # PEP-127) is available for use in neutralization and control experiments.

          Target Information

          Coatomer proteins are involved in regulating transport between the endoplasmic reticulum (ER) and the Golgi complex and in intra-Golgi transport. There exist two coatomer-protein mechanisms (COPI and COPII) and although they have mechanistic parallels, they are molecularly distinct. The COPI coat is comprised of seven subunits (alpha-, beta-, beta'-, gamma-, delta-, epsilon-, and zeta-COP) in a complex called coatomer. Assembly of the coatomer (COPI) onto non-clathrin coated vesicles is regulated by ADP-ribosylation factor (ARF). Vesicle formation, budding, fusion, and disassembly is dependent on GDP-GTP exchange, COPI, and ARF. COPI has been shown to facilitate retrograde intracellular transport from the ER to the Golgi complex. By contrast, COPII facilitates anterograde transport between these subcellular organelles. COPII has been shown to be independently and selectively recruited to the ER relative to COPI subunits. COPII consists of three parts: Sar1p and the two protein complexes, Sec23p-Sec24p and Sec13p-Sec31p.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ABP125; ABP130; cop ii; GATOR complex protein SEC13; GATOR2 complex protein SEC13; hSec23A; Protein SEC13 homolog; Protein transport protein Sec23A; Protein transport protein Sec24A; Protein transport protein Sec31A; SEC13 homolog, nuclear pore and COPII coating complex component; SEC13 related; SEC13-like 1; SEC13-like protein 1; SEC13-related protein; Sec23 homolog A, coat complex II component; SEC23-related protein A; SEC23A; SEC24 family, member A; SEC24 related gene family, member A; SEC24-related protein A; SEC31 homolog A, COPII coating complex component; SEC31-like 1; SEC31-like protein 1; SEC31-related protein A; unnamed protein product; vesicle associated protein; Vesicle-associated protein 1; Web1-like protein; yeast Sec31p homolog

          View more View less

          Gene Aliases: 1110003H02Rik; 1810024J13Rik; ABP125; ABP130; AU042160; CLSD; D3S1231E; HPBKS; hSec23A; HSPC275; HSPC334; KIAA0905; Msec23; NEDSOSB; npp-20; SEC13; SEC13A; SEC13L1; SEC13R; Sec23; SEC23A; Sec23r; SEC24A; SEC31A; SEC31L1; VAP1

          View more View less

          UniProt ID: (Human) P55735, (Human) Q15436, (Human) O95486, (Human) O94979, (Mouse) Q9D1M0, (Rat) Q5XFW8, (Mouse) Q01405, (Mouse) Q3U2P1, (Mouse) Q3UPL0, (Rat) Q9Z2Q1

          View more View less

          Entrez Gene ID: (Human) 6396, (Human) 10484, (Human) 10802, (Human) 22872, (Mouse) 110379, (Rat) 297522, (Mouse) 20334, (Rat) 58817, (Mouse) 77371, (Rat) 287275, (Mouse) 69162, (Rat) 93646

          View more View less

          Function(s)
          structural molecule activity protein binding identical protein binding GTPase activator activity zinc ion binding metal ion binding SNARE binding calcium-dependent protein binding molecular_function vesicle coat protein transporter
          Process(es)
          protein import into nucleus intracellular protein transport nucleocytoplasmic transport protein transport vesicle-mediated transport cellular response to nutrient levels positive regulation of TOR signaling protein exit from endoplasmic reticulum mRNA transport cargo loading into COPII-coated vesicle COPII-coated vesicle budding negative regulation of TORC1 signaling positive regulation of TORC1 signaling ER to Golgi vesicle-mediated transport protein localization to plasma membrane regulation of cholesterol transport cholesterol homeostasis positive regulation of protein secretion endoplasmic reticulum organization response to calcium ion transport regulation of low-density lipoprotein particle receptor biosynthetic process positive regulation of cholesterol homeostasis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline