Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HTR1B Antibodies

          Invitrogen

          HTR1B Polyclonal Antibody

          View all (13) HTR1B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HTR1B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          HTR1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HTR1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HTR1B Antibody (PA1-41069) in WB

          Western blot analysis of HTR1B in human brain lysate. Samples were incubated in HTR1B polyclonal antibody (Product # PA1-41069). A protein band of approximate molecular weight of 40-41 kDa was detected. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HTR1B Antibody in Western Blot (WB)
          HTR1B Polyclonal Antibody

          Product Details

          PA1-41069

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Mixture of synthetic peptides corresponding to amino acids 8-26 and 263-278 of rat 5-HT1BR.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2248464

          Product Specific Information

          Suggested positive control: human, mouse, or rat brain.

          Target Information

          Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. Molecular cloning studies have identified 5 receptors belonging to the 5-HT1 subtypes. These receptors are negatively coupled to adenylyl cyclase. Radioligand binding studies suggest the presence of 5-HT1B (formerly known as 5-HT1DbR) sites in substantia nigra, dorsal subiculum, globus pallidus, and superior collicum. In situ hybridization studies show abundance of 5-HT1BR mRNA in CA1 pyramidal cells of hippocampus, Purkinje cells of cerebellum, olfactory tubercle, subthalamic nucleus and somewhat lower levels in the caudate-putamen, cortical areas, thalamic and hypothalamic nuclei.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5-HT-1B; 5-HT-1D-beta; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT<1B> receptor; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; 5HT1B Receptor; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; Serotonin 1D beta receptor; Serotonin receptor 1B; unnamed protein product

          View more View less

          Gene Aliases: 5-HT-1B; 5-HT-1D-beta; 5-HT1B; 5-HT1DB; 5ht1b; HTR1B; HTR1D2; HTR1DB; S12

          View more View less

          UniProt ID: (Human) P28222, (Mouse) P28334, (Rat) P28564

          View more View less

          Entrez Gene ID: (Human) 3351, (Mouse) 15551, (Rat) 25075

          View more View less

          Function(s)
          Gi/o-coupled serotonin receptor activity G-protein coupled receptor activity G-protein coupled serotonin receptor activity protein binding neurotransmitter receptor activity serotonin binding serotonin receptor activity signal transducer activity drug binding G-protein coupled receptor
          Process(es)
          signal transduction G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting serotonin receptor signaling pathway phospholipase C-activating serotonin receptor signaling pathway chemical synaptic transmission negative regulation of serotonin secretion vasoconstriction bone remodeling regulation of behavior cellular response to alkaloid cellular response to xenobiotic stimulus positive regulation of vascular smooth muscle cell proliferation G-protein coupled receptor internalization protein kinase C-activating G-protein coupled receptor signaling pathway serotonin receptor signaling pathway behavior feeding behavior response to toxic substance regulation of dopamine secretion negative regulation of cAMP biosynthetic process negative regulation of synaptic transmission, GABAergic cellular response to drug response to cocaine response to drug drinking behavior response to ethanol negative regulation of synaptic transmission, glutamatergic cellular response to temperature stimulus response to mineralocorticoid
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline