Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
This antibody is predicted to react with xenopus based on sequence homology.
Immunogen sequence homology: Gorilla (94%).
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor 72; G-protein coupled receptor 83; glucocorticoid induced receptor; Glucocorticoid-induced receptor; Hcepto9; Jp05; probable G-protein coupled receptor 83; unnamed protein product
Gene Aliases: AW045253; GIR; GPR72; GPR83; JP05; KIAA1540; RP105; RP39; RP82
UniProt ID: (Dog) Q9TTQ9, (Human) Q9NYM4, (Mouse) P30731, (Rat) Q8VHD7
Entrez Gene ID: (Dog) 403699, (Human) 10888, (Chicken) 428093, (Mouse) 14608, (Rat) 140595
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support