Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PMP70 Antibodies

          Invitrogen

          PMP70 Polyclonal Antibody, Alexa Fluor™ 555

          1 Reference
          View all (15) PMP70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PMP70 Polyclonal Antibody, Alexa Fluor™ 555

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PMP70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PMP70 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PMP70 Antibody (PA1-650-A555) in ICC/IF

          Immunofluorescent analysis of PMP-70 (orange) in HeLa cells. The cells were fixed with 4% Paraformaldehyde in PBS for 15 minutes at room temperature, and blocked with 3% BSA in PBS (Product # 37525) for 30 minutes at room temperature. Cells were stained with a PMP-70 Polyclonal Antibody, AlexaFluor 555 conjugate (Product # PA1-650-A555) at a dilution of 2.5 µg/mL in blocking buffer for 1 hour at room temperature protected from light. Nuclei (blue) were stained with Hoechst Dye (Product # 62249) at a dilution of 1:10,000 in blocking buffer.... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PMP70 Antibody in Immunocytochemistry (ICC/IF)
          PMP70 Polyclonal Antibody, Alexa Fluor™ 555

          Product Details

          PA1-650-A555

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1-10 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic Peptide: C N(644) Y E F K K I T E D T V E F G S(659)
          3D Epitope / Immunogen

          Conjugate

          Alexa Fluor™ 555 Alexa Fluor™ 555 Alexa Fluor™ 555

          Excitation/Emission Max

          553/568 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2662745

          Product Specific Information

          PA1650A555 immunizing peptide corresponds to amino acid residues 644-659 from rat PMP70. This sequence is completely conserved in mouse PMP70 and differs from human PMP70 by a single amino acid substitution. The immunizing peptide (Cat. # PEP-038) is available for use in neutralization and control experiments.

          Target Information

          Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum" disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ATP-binding cassette sub-family D member 3; ATP-binding cassette, sub-family D (ALD), member 3; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein 70 kDa; peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; PMP70; unnamed protein product

          View more View less

          Gene Aliases: ABC43; ABCD3; AI313901; AU018866; AW146054; CBAS5; PMP68; PMP70; PXMP1; ZWS2

          View more View less

          UniProt ID: (Human) P28288, (Mouse) P55096, (Rat) P16970

          View more View less

          Entrez Gene ID: (Human) 5825, (Mouse) 19299, (Rat) 25270

          View more View less

          Function(s)
          nucleotide binding long-chain fatty acid transporter activity protein binding ATP binding hydrolase activity ATPase activity ATPase activity, coupled to transmembrane movement of substances identical protein binding protein homodimerization activity acyl-CoA hydrolase activity ABC-type transporter activity protein self-association ATP-binding cassette (ABC) transporter
          Process(es)
          very long-chain fatty acid metabolic process fatty acid biosynthetic process fatty acid beta-oxidation bile acid biosynthetic process lipid transport peroxisome organization response to xenobiotic stimulus bile acid and bile salt transport peroxisomal long-chain fatty acid import very long-chain fatty acid catabolic process transmembrane transport phytanic acid metabolic process transport metabolic process response to organic cyclic compound response to drug
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline