Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Dynamin 2 Antibodies

          Invitrogen

          Dynamin 2 Polyclonal Antibody

          6 Published Figures
          20 References
          View all (15) Dynamin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Dynamin 2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (6)
          Dynamin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Dynamin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Dynamin 2 Antibody (PA1-661) in ICC/IF

          Immunofluorescence analysis of Dynamin 2 was performed using 70% confluent log phase SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Dynamin 2 Polyclonal Antibody (Product # PA1-661) at 1:100 in 0.1% BSA, incubated at 4 degree celsius over... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Dynamin 2 Antibody in Immunocytochemistry (ICC/IF)
          Dynamin 2 Antibody in Immunocytochemistry (ICC/IF)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Antibody in Immunohistochemistry (IHC)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Antibody in Western Blot (WB)
          Dynamin 2 Polyclonal Antibody

          Product Details

          PA1-661

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2-4 µg/mL
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic Peptide: C S(760) P T P Q R R P V S S I H P P G R P P A(779)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2293040

          Product Specific Information

          PA1-661 detects dynamin II (Dyn2) from rat, mouse, and human tissues and cells.

          PA1-661 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~100 kDa protein representing Dyn2 from HeLa cell lysate.

          PA1-661 immunizing peptide corresponds to amino acid residues 760-779 from human Dyn2. This sequence is 95% conserved in mouse and rat Dyn2. PA1-661 immunizing peptide (Cat. # PEP-056) is available for use in neutralization and control experiments.

          Target Information

          Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Five alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Dynamin 2; Dynamin II; Dynamin UDNM; Dynamin-2; unnamed protein product

          View more View less

          Gene Aliases: b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYIIAAB; DYN2; DYNII; LCCS5; Udnm

          View more View less

          UniProt ID: (Human) P50570, (Rat) P39052, (Mouse) P39054

          View more View less

          Entrez Gene ID: (Human) 1785, (Rat) 25751, (Mouse) 13430

          View more View less

          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding phosphatidylinositol-4,5-bisphosphate binding microtubule binding hydrolase activity SH3 domain binding enzyme binding protein kinase binding D2 dopamine receptor binding protein complex binding phosphatidylinositol 3-kinase regulatory subunit binding WW domain binding nitric-oxide synthase binding membrane traffic protein
          Process(es)
          G2/M transition of mitotic cell cycle regulation of transcription, DNA-templated post-Golgi vesicle-mediated transport endocytosis receptor-mediated endocytosis phagocytosis autophagy centrosome cycle signal transduction synaptic vesicle budding from presynaptic endocytic zone membrane antigen processing and presentation of exogenous peptide antigen via MHC class II regulation of axon extension receptor internalization transferrin transport positive regulation of apoptotic process stress fiber assembly positive regulation of transcription, DNA-templated synaptic vesicle transport neuron projection morphogenesis protein polymerization membrane organization actin filament bundle organization membrane tubulation vesicle scission negative regulation of membrane tubulation G-protein coupled receptor internalization Golgi to plasma membrane transport spermatogenesis metabolic process response to light stimulus positive regulation of lamellipodium assembly negative regulation of transforming growth factor beta receptor signaling pathway regulation of Rac protein signal transduction response to cocaine macropinocytosis positive regulation of nitric oxide biosynthetic process positive regulation of endocytosis positive regulation of phagocytosis cilium morphogenesis cellular response to carbon monoxide cellular response to X-ray cellular response to nitric oxide positive regulation of substrate adhesion-dependent cell spreading cellular response to dopamine regulation of Golgi organization positive regulation of sodium:potassium-exchanging ATPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline