Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
PA1-759 detects ubiquilin from hamster cells.
PA1-759 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~63 kDa protein representing ubiquilin from CHO cell extract.
PA1-759 immunizing peptide corresponds to amino acids residues 2-18 from mouse ubiquilin.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer's and Parkinson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DSK2 homolog; hPLIC-1; PLIC-1; Protein linking IAP with cytoskeleton 1; similar to yeast DSK2; testicular tissue protein Li 219; ubiquilin 1; Ubiquilin-1; ubiquitin-like type II; unnamed protein product
Gene Aliases: 1110046H03Rik; 1810030E05Rik; AU019746; C77538; D13Ertd372e; DA41; DSK2; I79_020705; PLIC-1; PLIC1; UBQLN1; UBQN; XDRP1
UniProt ID: (Human) Q9UMX0, (Mouse) Q8R317
Entrez Gene ID: (Human) 29979, (Mouse) 56085, (Chinese hamster) 100764107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support