Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HSP90 alpha Antibodies

          Invitrogen

          HSP90 alpha Polyclonal Antibody

          Advanced Verification
          32 Published Figures
          53 References
          View all (33) HSP90 alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HSP90 alpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          • Published Figures (32)
          • Advanced Verification (1)
          HSP90 alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HSP90 alpha Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HSP90 alpha Antibody (PA3-013) in ICC/IF

          Immunofluorescent analysis of Heat Shock Protein 86 (HSP86, green) in HeLa cells and NIH3T3 cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature and blocked with 1% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were probed with a HSP86 polyclonal antibody (Product # PA3-013), at a dilution of 1:100 for at least 1 hour at ro... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HSP90 alpha Antibody in Immunocytochemistry (ICC/IF)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSP90 alpha Antibody in Immunoprecipitation (IP)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Immunocytochemistry (ICC/IF)
          HSP90 alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSP90 alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSP90 alpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Immunocytochemistry (ICC/IF)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot, Immunoprecipitation (WB, IP)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot, Immunoprecipitation (WB, IP)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Western Blot, Immunoprecipitation (WB, IP)
          HSP90 alpha Antibody in Western Blot (WB)
          HSP90 alpha Antibody in Immunoprecipitation (IP)
          HSP90 alpha Antibody in Immunoprecipitation (IP)
          HSP90 alpha Antibody
          HSP90 alpha Polyclonal Antibody

          Product Details

          PA3-013

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 37 publications 37 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          2 µg
          View 4 publications 4 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Sheep, Rat

          Published species

          Human, Mouse, Non-human primate, Rabbit

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues P(2) E E T Q T Q D Q P M(12) of mouse HSP86.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2120934

          Product Specific Information

          PA3-013 detects heat shock protein 90 (HSP90) from human, monkey, mouse, sheep (adrenal) and rat tissues and cells. This antibody does not detect HSP84.

          PA3-013 has been successfully used in Western blot, immunofluorescence, immunohistochemistry (paraffin) and immunoprecipitation procedures. By Western blot, this antibody detects an ~86 kDa protein representing HSP86 from HeLa cell lysate. Immunofluorescence staining of HSP86 in HeLa cells with PA3-013 yields a pattern consistent with cytoplasmic staining. Immunoprecipitation experiments with this antibody suggest that HSP86 exists primarily as homodimers in HeLa cells. Furthermore, the antibody is capable of precipitating HSP86 that is complexed with other proteins such as the aryl hydrocarbon (Ah) receptor.

          The PA3-013 immunogen is a synthetic peptide corresponding to residues P(2) E E T Q T Q D Q P M(12) of mouse HSP86. The N-terminal regions of HSP84 and HSP86 show the largest difference in amino acid sequence.

          Target Information

          Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; Heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein family C member 1; Heat shock protein HSP 90-alpha; heat shock protein, 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; Heat shock protein90; HSP; HSP 86; HSP 90 alpha; HSP86; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; Renal carcinoma antigen NY-REN-38; TSTA; Tumor-specific transplantation 86 kDa antigen; unnamed protein product

          View more View less

          Gene Aliases: 86kDa; 89kDa; AL024080; AL024147; EL52; HEL-S-65p; Hsp103; hsp4; HSP86; Hsp86-1; Hsp89; HSP89A; Hsp90; HSP90A; HSP90AA1; HSP90N; HSPC1; HSPCA; HSPCAL1; HSPCAL4; HSPN; LAP-2; LAP2

          View more View less

          UniProt ID: (Human) P07900, (Mouse) P07901, (Rat) P82995

          View more View less

          Entrez Gene ID: (Sheep) 100127209, (Human) 3320, (Mouse) 15519, (Rat) 299331

          View more View less

          Function(s)
          nucleotide binding UTP binding CTP binding RNA binding mRNA binding protein binding ATP binding GTP binding hydrolase activity ATPase activity sulfonylurea receptor binding protein phosphatase binding MHC class II protein complex binding nitric-oxide synthase regulator activity TPR domain binding ubiquitin protein ligase binding dATP binding identical protein binding protein homodimerization activity histone deacetylase binding protein binding involved in protein folding ion channel binding tau protein binding GTPase binding Rho GDP-dissociation inhibitor binding unfolded protein binding DNA polymerase binding scaffold protein binding disordered domain specific binding ATP-dependent protein folding chaperone enzyme-substrate adaptor activity protein tyrosine kinase binding protein kinase binding poly(A) RNA binding Hsp90 family chaperone
          Process(es)
          neuron migration activation of innate immune response positive regulation of defense response to virus by host skeletal muscle contraction protein folding mitochondrial transport response to unfolded protein telomere maintenance via telomerase response to heat response to cold response to xenobiotic stimulus response to salt stress positive regulation of lamellipodium assembly cardiac muscle cell apoptotic process protein import into mitochondrial matrix regulation of protein ubiquitination positive regulation of telomere maintenance via telomerase positive regulation of protein polymerization positive regulation of interferon-beta production regulation of protein localization cellular response to heat protein refolding response to cocaine positive regulation of protein import into nucleus regulation of apoptotic process regulation of protein complex assembly protein unfolding response to estrogen positive regulation of nitric oxide biosynthetic process positive regulation of protein catabolic process positive regulation of cell size nitric oxide metabolic process response to antibiotic protein stabilization chaperone-mediated protein complex assembly positive regulation of cardiac muscle contraction chaperone-mediated autophagy cellular response to virus regulation of postsynaptic membrane neurotransmitter receptor levels neurofibrillary tangle assembly telomerase holoenzyme complex assembly nitric oxide biosynthetic process response to stress positive regulation of protein import into nucleus, translocation protein import into mitochondrial outer membrane positive regulation of cytotoxic T cell differentiation regulation of catalytic activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline