Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
PA3-067 detects Endothelin 1 from human, mouse, and rat samples.
PA3-067 has been successfully used in Western blot, immunocytochemistry, and immunohistochemistry procedures. By Western blot, this antibody detects a 24.4 kDa protein corresponding to mouse Endothelin 1.
The PA3-067 antigen is mouse Endothelin-1 CS-Abu-SSLMDKE-Abu-VYF-Abu-HLDIIW coupled to KLH.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Big endothelin 1; Big ET; Big ET 1; Big ET1; BigET; EDN1; Endothelin-1; Et; ET-1; PPET1; preproendothelin; Preproendothelin-1; unnamed protein product
Gene Aliases: ARCND3; EDN1; ET-1; ET1; HDLCQ7; PPET1; preproET; QME
UniProt ID: (Human) P05305, (Rat) P22388, (Mouse) P22387
Entrez Gene ID: (Human) 1906, (Rat) 24323, (Mouse) 13614
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support