Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • SR-BI/SR-BII Antibodies

          Invitrogen

          SR-BI/SR-BII Polyclonal Antibody

          2 References
          View all (5) SR-BI/SR-BII antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SR-BI/SR-BII Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          SR-BI/SR-BII Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SR-BI/SR-BII Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SR-BI/SR-BII Antibody (PA3-16805) in ICC/IF

          Immunocytochemistry analysis of SR-BI/SR-BII in HeLa cells. Samples were incubated with SR-BI/SR-BII polyclonal antibody (Product # PA3-16805) followed by DyLight 488 (green). Nuclei and alpha-tubulin were counterstained with DAPI (blue) and Dylight 550 (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SR-BI/SR-BII Antibody in Immunocytochemistry (ICC/IF)
          SR-BI/SR-BII Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SR-BI/SR-BII Antibody in Immunohistochemistry (IHC)
          SR-BI/SR-BII Antibody in Western Blot (WB)
          SR-BI/SR-BII Antibody in Flow Cytometry (Flow)
          SR-BI/SR-BII Polyclonal Antibody

          Product Details

          PA3-16805

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:1,000
          -

          Flow Cytometry (Flow)

          1:400
          -

          Immunoprecipitation (IP)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2182692

          Target Information

          High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. SR-BI and SR-BII (previously known as SR-BI.2) are the alternatively spliced products of a single gene. SR-BII and SR-BI are identical except for the encoded c-terminal cytoplasmic domain. Both SR-BI and SR-BII bind HDL and mediates selective uptake of HDL cholesteryl ester, but with SR-BII having an approximately 4-fold lower efficiency than SR-BI. SR-BI and SR-BII are expressed primarily in liver and non-placental steroidgenic tissues. Although the role of these scavenger receptors is not completely clear, SR-BII mRNA results from the alternative splicing of SR-BI precursor transcripts with both isoforms mediating selective transfer of lipid between HDL and cells. Therefore, the relative expression and functional activities of these two isoforms create a potential means of regulating selective lipid transfer between HDL and cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 1; CD36 antigen-like 2; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; HDL QTL 1; LGP85; LIMP II; Lysosome membrane protein 2; Lysosome membrane protein II; scavanger receptor class B type 1; scavenger receptor b1; Scavenger receptor class B member 1; Scavenger receptor class B member 2; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B1; SR-B1/B2; SR-BI; SR-BI/BII; SRB1; unnamed protein product

          View more View less

          Gene Aliases: 9330185J12Rik; AI120173; AMRF; CD36; CD36L1; CD36L2; CLA-1; CLA1; D5Ertd460e; EPM4; HDLCQ6; Hdlq1; HDLQTL6; Hlb398; HLGP85; LGP85; LIMP-2; LIMP2; LIMPII; MLGP85; mSR-BI; SCARB1; SCARB2; SR-B1; SR-BI; SR-BII; SRB1; SRBI

          View more View less

          UniProt ID: (Human) Q8WTV0, (Human) Q14108, (Rat) P97943, (Mouse) Q61009, (Rat) P27615, (Mouse) O35114

          View more View less

          Entrez Gene ID: (Human) 949, (Human) 950, (Rat) 25073, (Mouse) 20778, (Rat) 117106, (Mouse) 12492

          View more View less

          Function(s)
          lipopolysaccharide binding beta-amyloid binding virus receptor activity phosphatidylserine binding lipopolysaccharide receptor activity scavenger receptor activity protein binding 1-phosphatidylinositol binding high-density lipoprotein particle binding lipid binding low-density lipoprotein particle binding apolipoprotein binding apolipoprotein A-I binding high-density lipoprotein particle receptor activity transmembrane signaling receptor activity cholesterol binding enzyme binding phosphatidylcholine binding cargo receptor activity protein homodimerization activity chaperone binding lipid transporter activity receptor activity membrane trafficking regulatory protein
          Process(es)
          endothelial cell proliferation cholesterol catabolic process lipid transport positive regulation of endothelial cell migration positive regulation of nitric oxide mediated signal transduction positive regulation of triglyceride biosynthetic process positive regulation of cholesterol storage regulation of phosphatidylcholine catabolic process phospholipid transport lipopolysaccharide transport cholesterol transport lipopolysaccharide-mediated signaling pathway detection of lipopolysaccharide cholesterol efflux high-density lipoprotein particle remodeling plasma lipoprotein particle clearance low-density lipoprotein particle clearance high-density lipoprotein particle clearance vitamin transmembrane transport wound healing vasodilation cholesterol homeostasis blood vessel endothelial cell migration recognition of apoptotic cell reverse cholesterol transport adhesion of symbiont to host viral entry into host cell regulation of phagocytosis high density lipoprotein particle mediated signaling triglyceride homeostasis cholesterol import intestinal lipid absorption positive regulation of sphingolipid mediated signaling pathway protein targeting to lysosome receptor-mediated endocytosis sensory perception of sound positive regulation of neuron projection development aminophospholipid transport regulation of carbohydrate catabolic process endosome to plasma membrane protein transport regulation of endosome organization regulation of lysosome organization regulation of glucosylceramide catabolic process steroid biosynthetic process androgen biosynthetic process phagocytosis, recognition cell adhesion positive regulation of nitric-oxide synthase activity pattern recognition receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline