Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Calreticulin Antibodies

          Invitrogen

          Calreticulin Polyclonal Antibody

          81 Published Figures
          260 References
          View all (42) Calreticulin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calreticulin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          • Published Figures (81)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 93

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calreticulin Antibody (PA3-900) in ICC/IF

          Immunofluorescent analysis of Calreticulin using anti-Calreticulin polyclonal antibody (Product # PA3-900) shows staining in NS-1 Cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunohistochemistry (IHC)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry, Immunoprecipitation (ICC/IF, IP)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunocytochemistry (ICC/IF)
          Calreticulin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Calreticulin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Calreticulin Antibody in Western Blot, Immunohistochemistry, Immunoprecipitation (WB, IHC, IP)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot (WB)
          Calreticulin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)
          Calreticulin Antibody in Immunoprecipitation (IP)

          Product Details

          PA3-900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 116 publications 116 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 18 publications 18 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:50 - 1:200
          View 81 publications 81 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 17 publications 17 publications

          ELISA (ELISA)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 21 publications 21 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate, Rabbit, Rat

          Published species

          Amphibian, Bacteria, Bovine, Dog, Echinoderm, Hamster, Human, Mouse, Non-human primate, Rat, Virus, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant, human calreticulin protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325990

          Product Specific Information

          PA3-900 detects calreticulin from human, non-human primate, rabbit, mouse, canine and rat samples.



          PA3-900 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, flow cytometry, and immunoprecipitation procedures. By Western blot, this antibody detects an ~60 kDa protein representing calreticulin from rat brain extract. Immunofluorescence staining of calreticulin in rat brain cortex with PA3-900 results in fine granular staining consistent with ER localization.



          PA3-900 immunogen is recombinant human calreticulin protein produced in the Baculovirus insect cell system.

          Target Information

          Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Autoantigen RO; CABP3; CALBP; Calcium-binding protein 3; CALR; Calregulin; Calreticulin; calreticulin precursor; CRP55; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; Ro ribonucleoprotein autoantigen (Ro/SS-A) precursor; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); unnamed protein product

          View more View less

          Gene Aliases: CALR; CALR1; Calregulin; cC1qR; CRP55; CRT; CRTC; ERp60; HACBP; HEL-S-99n; RO; SSA

          View more View less

          UniProt ID: (Human) P27797, (Rabbit) P15253, (Mouse) P14211, (Rat) P18418

          View more View less

          Entrez Gene ID: (Rabbit) 100350417, (Human) 811, (Rabbit) 100009050, (Dog) 476694, (Mouse) 12317, (Rat) 64202

          View more View less

          Function(s)
          complement component C1q binding DNA binding RNA binding mRNA binding nuclear export signal receptor activity integrin binding iron ion binding calcium ion binding protein binding zinc ion binding carbohydrate binding ubiquitin protein ligase binding peptide binding hormone binding protein binding involved in protein folding metal ion binding androgen receptor binding unfolded protein binding chaperone binding molecular sequestering activity glycoprotein binding poly(A) RNA binding chaperone
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter peptide antigen assembly with MHC class I protein complex regulation of transcription, DNA-templated protein folding protein export from nucleus cellular calcium ion homeostasis spermatogenesis positive regulation of cell proliferation response to xenobiotic stimulus positive regulation of endothelial cell migration positive regulation of gene expression negative regulation of translation cortical actin cytoskeleton organization response to estradiol negative regulation of intracellular steroid hormone receptor signaling pathway response to testosterone protein localization to nucleus protein folding in endoplasmic reticulum ERAD pathway regulation of meiotic nuclear division glucocorticoid receptor signaling pathway regulation of apoptotic process negative regulation of neuron differentiation positive regulation of cell cycle negative regulation of transcription, DNA-templated negative regulation of retinoic acid receptor signaling pathway positive regulation of phagocytosis protein stabilization sequestering of calcium ion protein maturation cardiac muscle cell differentiation cellular response to electrical stimulus cellular response to lithium ion cellular senescence cellular response to virus positive regulation of substrate adhesion-dependent cell spreading negative regulation of trophoblast cell migration positive regulation of NIK/NF-kappaB signaling response to peptide response to glycoside response to biphenyl positive regulation of dendritic cell chemotaxis cell cycle arrest positive regulation of DNA replication chaperone-mediated protein folding response to organic substance response to drug cellular response to organic substance
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline