Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • AEBP2 Antibodies

          Invitrogen

          AEBP2 Polyclonal Antibody

          View all (16) AEBP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AEBP2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (29)
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 29

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AEBP2 Antibody (PA5-100088) in IHC (P)

          Immunohistochemistry analysis of AEBP2 in human normal tissues adjacent to colorectal cancer. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with AEBP2 polyclonal antibody (Product # PA5-100088) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AEBP2 Antibody in Western Blot (WB)
          AEBP2 Antibody in Western Blot (WB)
          AEBP2 Polyclonal Antibody

          Product Details

          PA5-100088

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human AEBP2(Accession Q6ZN18), corresponding to amino acid residues H352-T402.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2815618

          Product Specific Information

          Antibody detects endogenous levels of total AEBP2.

          Target Information

          AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates 'Lys-9' and 'Lys-27' residues of histone H3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Adipocyte enhancer-binding protein 2; AE(adipocyte enhancer)-binding protein 2; AE-binding protein 2; unnamed protein product; Zinc finger protein AEBP2

          View more View less

          Gene Aliases: AEBP2; AU023766; B230313N05Rik

          View more View less

          UniProt ID: (Human) Q6ZN18, (Mouse) Q9Z248

          View more View less

          Entrez Gene ID: (Human) 121536, (Rat) 297705, (Mouse) 11569

          View more View less

          Function(s)
          DNA binding transcription cofactor activity protein binding zinc ion binding metal ion binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding transcription corepressor activity nucleic acid binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter chromatin organization regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter transcription, DNA-templated chromatin modification
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline