Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Antibody detects endogenous levels of total ARHGAP15.
RHO GTPases regulate diverse biologic processes, and their activity is regulated by RHO GTPase-activating proteins (GAPs), such as ARHGAP15 (Seoh et al., 2003 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ArhGAP15; Rho GTPase-activating protein 15; Rho-type GTPase-activating protein 15; RhoGAP; RacGAP protein; unnamed protein product
Gene Aliases: 5830480G12Rik; ARHGAP15; BM-024; BM-030; BM-046; BM046
UniProt ID: (Human) Q53QZ3, (Rat) Q6AYC5, (Mouse) Q3T9M8
Entrez Gene ID: (Human) 55843, (Rat) 295635, (Mouse) 76117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support