Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total CNOT6.
Poly(A) nuclease with 3'-5' RNase activity. Catalytic component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Involved in mRNA decay mediated by the major-protein-coding determinant of instability (mCRD) of the FOS gene in the cytoplasm. In the presence of ZNF335, enhances ligand-dependent transcriptional activity of nuclear hormone receptors, including RARA. The increase of ligand-dependent ESR1-mediated transcription is much smaller, if any. Mediates cell proliferation and cell survival and prevents cellular senescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbon catabolite repression 4 protein; Carbon catabolite repressor protein 4 homolog; CCR4 carbon catabolite repression 4-like; CCR4-NOT transcription complex subunit 6; Cytoplasmic deadenylase; unnamed protein product
Gene Aliases: CCR4; CCR4a; CNOT6; KIAA1194; RGD1310783
UniProt ID: (Human) Q9ULM6, (Rat) Q6AXU9, (Mouse) Q8K3P5
Entrez Gene ID: (Human) 57472, (Rat) 287249, (Mouse) 104625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support