Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total MED20.
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTRFP; Mediator complex subunit 20; Mediator of RNA polymerase II transcription subunit 20; predicted protein of HQ0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRF-proximal protein homolog; TRFP; RNA polymerase II holoenzyme component; ubiquitin specific protease 49; unnamed protein product
Gene Aliases: 1110011O05Rik; 2410115I17Rik; AU018348; MED20; PRO0213; SRB2; TRFP; Usp49
UniProt ID: (Human) Q9H944, (Mouse) Q9R0X0, (Rat) Q5XIE9
Entrez Gene ID: (Human) 9477, (Mouse) 56771, (Rat) 316209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support