Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total PKNOX2.
PREP-2 (Pbx-regulating protein-2), also known as PBX/knotted 1 homeobox 2 or PKNOX2, is a widely expressed protein belonging to the TALE (three amino acid loop extension)/MEIS family. PREP-2 is a DNA-binding protein that forms stable complexes with Pbx proteins. It is highly homologous to the related protein PREP-1, but displays a more restricted tissue distribution and a higher DNA-dissociation rate. Like PREP-1, PREP-2 forms a heterodimer with Pbx 1. The PREP-2-Pbx 1 dimer is relocated to the nucleus where it associates with HoxB1 to form a ternary complex. In contrast with PREP-1, which acts to increase transcriptional activation in this ternary complex, PREP-2 leads to a slight decrease in transcriptional activity of the ternary complex. Multiple isoforms exist for PREP-2, localizing to the nucleus or cytoplasm. Cytoplasmic isoforms are believed to colocalize with F-actin, G-actin and tubulin/microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein PKNOX2; Homeobox protein PREP-2; homeodomain containing transcription factor PREP2; PBX/knotted homeobox 2; unnamed protein product
Gene Aliases: D230005H23Rik; PKNOX2; PREP2
UniProt ID: (Human) Q96KN3, (Mouse) Q8BG99
Entrez Gene ID: (Human) 63876, (Mouse) 208076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support