Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CYP1A1/CYP1A2 Antibodies

          Invitrogen

          CYP1A1/CYP1A2 Polyclonal Antibody

          View all (5) CYP1A1/CYP1A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CYP1A1/CYP1A2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CYP1A1/CYP1A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CYP1A1/CYP1A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CYP1A1/CYP1A2 Antibody (PA5-101307) in ICC/IF

          Immunofluorescence analysis of Cytochrome P450 1A1/A2 was performed using T-47D cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with CYP1A1 Polyclonal Antibody (Product # PA5-101307) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnigh... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CYP1A1/CYP1A2 Antibody in Immunocytochemistry (ICC/IF)
          CYP1A1/CYP1A2 Antibody in Western Blot (WB)
          CYP1A1/CYP1A2 Antibody in Western Blot (WB)

          Product Details

          PA5-101307

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CYP1A1(Accession P04798), corresponding to amino acid residues I78-G128.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2815888

          Product Specific Information

          Antibody detects endogenous levels of total Cytochrome P450 1A1/2.

          Target Information

          This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. Other xenobiotic substrates for this enzyme include caffeine, aflatoxin B1, and acetaminophen. The transcript from this gene contains four Alu sequences flanked by direct repeats in the 3' untranslated region.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: aromatic compound inducible; aryl hydrocarbon hydroxylase; Cholesterol 25-hydroxylase; CYP1A1; CYP1A1v; alternatively spliced; CYPIA1; CYPIA2; Cytochrome P(3)450; Cytochrome P-448; cytochrome P-450 peptide; cytochrome P-450 protein; cytochrome P-450-1; Cytochrome P-450d; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P3 450; Cytochrome P450 1A1; Cytochrome P450 1A2; Cytochrome P450 4; cytochrome P450 family 1 subfamily a polypeptide 1; cytochrome P450 family 1 subfamily a polypeptide 2; Cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily A, polypeptide 1; cytochrome P450, family 1, subfamily A, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; Cytochrome P450-C; Cytochrome P450-D; Cytochrome P450-P1; Cytochrome P450-P2; Cytochrome P450-P3; Cytochrome P450MT2; dioxin-inducible; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; functional drug metabolizing enzyme; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P-448; P450 form 4; P450-D; P450form6; P450MT2; unnamed protein product; xenobiotic monooxygenase

          View more View less

          Gene Aliases: AHH; AHRR; CP11; CP12; CYP1; Cyp1a-1; Cyp1a-2; CYP1A1; CYP1A2; Cyp45c; Cypc45c; CYPD45; CYPIA1; CYPIA2; P-450d; P-450MC; P1-450; P3-450; P450(PA); P450-1; P450-3; P450-C; P450DX; RATCYPD45

          View more View less

          UniProt ID: (Human) P04798, (Human) P05177, (Mouse) P00184, (Rat) P00185, (Mouse) P00186, (Rat) Q64588

          View more View less

          Entrez Gene ID: (Human) 1543, (Human) 1544, (Mouse) 13076, (Rat) 24296, (Mouse) 13077, (Rat) 24297

          View more View less

          Function(s)
          catalytic activity monooxygenase activity iron ion binding protein binding arachidonic acid monooxygenase activity steroid hydroxylase activity oxidoreductase activity oxidoreductase activity, acting on diphenols and related substances as donors oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen flavonoid 3'-monooxygenase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen lyase activity oxygen binding enzyme binding heme binding Hsp70 protein binding demethylase activity metal ion binding Hsp90 protein binding vitamin D 24-hydroxylase activity estrogen 16-alpha-hydroxylase activity estrogen 2-hydroxylase activity long-chain fatty acid omega-hydroxylase activity hydroperoxy icosatetraenoate dehydratase activity long-chain fatty acid omega-1 hydroxylase activity electron carrier activity caffeine oxidase activity aromatase activity
          Process(es)
          response to hypoxia long-chain fatty acid metabolic process liver development lipid hydroxylation lipid metabolic process fatty acid metabolic process steroid biosynthetic process steroid catabolic process porphyrin-containing compound metabolic process xenobiotic metabolic process steroid metabolic process estrogen metabolic process amine metabolic process toxin metabolic process response to xenobiotic stimulus response to nematode response to herbicide response to toxic substance ethylene metabolic process coumarin metabolic process flavonoid metabolic process response to iron(III) ion insecticide metabolic process dibenzo-p-dioxin metabolic process phenol-containing compound metabolic process dibenzo-p-dioxin catabolic process epoxygenase P450 pathway response to food response to lipopolysaccharide response to vitamin A response to genistein response to immobilization stress xenobiotic catabolic process vitamin D metabolic process hormone metabolic process retinol metabolic process long-chain fatty acid biosynthetic process 9-cis-retinoic acid biosynthetic process camera-type eye development nitric oxide metabolic process response to arsenic-containing substance digestive tract development tissue remodeling hydrogen peroxide biosynthetic process response to hyperoxia maternal process involved in parturition hepatocyte differentiation cellular response to copper ion omega-hydroxylase P450 pathway olefinic compound metabolic process positive regulation of G1/S transition of mitotic cell cycle response to diphenyl ether response to Aroclor 1254 response to 2,3,7,8-tetrachlorodibenzodioxine response to 3-methylcholanthrene drug metabolic process heterocycle metabolic process oxidation-reduction process demethylation cellular response to organic cyclic compound cellular aromatic compound metabolic process toxin biosynthetic process post-embryonic development alkaloid metabolic process regulation of gene expression monoterpenoid metabolic process lung development monocarboxylic acid metabolic process drug catabolic process exogenous drug catabolic process cellular respiration oxidative demethylation cellular response to cadmium ion oxidative deethylation aging cell proliferation response to wounding response to virus embryo development ending in birth or egg hatching response to organic substance response to organic cyclic compound developmental process response to drug response to antibiotic cholesterol metabolic process arachidonic acid metabolic process methylation aflatoxin metabolic process response to estradiol
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline