Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total ALS2CR11.
ALS2CR11 has 2 isoforms of the human protein that are produced by alternative splicing. An autosomal recessive form of juvenile amyotrophic lateral sclerosis was originally mapped to a region of chromosome 2 that includes this gene. The encoded protein contains a calcium-dependent membrane targeting C2 domain. This domain is often found in proteins that are involved in membrane trafficking and signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALS2CR11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 11; C2 calcium dependent domain containing 6; C2 calcium-dependent domain-containing 6; C2 calcium-dependent domain-containing protein 6; Cation channel sperm-associated targeting subunit tau; Catsper channel auxiliary subunit tau; CatSper-tau; testicular tissue protein Li 16; unnamed protein product
Gene Aliases: 1700052H20Rik; 4930408G06Rik; ALS2CR11; C2CD6; CATSPERT; SPGF68
UniProt ID: (Mouse) A0A5F8MPU3, (Human) Q53TS8
Entrez Gene ID: (Mouse) 73463, (Human) 151254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support