Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total KIR2DL3.
NKAT (NK-associated transcripts) gene products, known as killer immuno-globulin-like receptors or KIRs, downregulate the cytotoxicity of NK cells upon recognition of specific class I major histocompatibility complex (MHC) molecules on target cells. This family of receptors is characterized by an extracellular region with two to three immunoglobulin-superfamily domains and a cytoplasmic domain with an antigen receptor activation motif (ARAM). KIRs and other inhibitory receptors also possess a common cytoplasmic sequence (I/VxYxxL/V) known as an ITIM (immunoreceptor tyrosine-based inhibitory motif). The human inhibitory human killer cell immunoglobulin-like receptor 2DL3 (KIR2DL3), also referred to as CD158b, is an inhibitory receptor that is specific for the human MHC class I molecule HLA-Cw3 and related alleles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD158 antigen-like family member B2; CD158b2; CU464054.1; extracellular domain; Killer cell immunoglobulin-like receptor 2DL3; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 3; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; Killer inhibitory receptor cl 2-3; KIR-023GB; MGC129943; natural killer associated transcript 2; natural killer cell inhibitory receptor KIR2DL3; natural killer cell receptor; Natural killer-associated transcript 2; NKAT-2; NKAT2a; nkat2a; putative; NKAT2b; nkat2b; putative; p58 KIR; p58 natural killer cell receptor clone CL-6; p58 NK receptor CL-6; p58.2 MHC class-I specific NK receptor; p58.2 MHC class-I-specific NK receptor
Gene Aliases: CD158b; CD158B2; GL183; KIR-023GB; KIR-K7b; KIR-K7c; KIR2DL; KIR2DL3; KIR2DS5; KIRCL23; NKAT; NKAT2; NKAT2A; NKAT2B; p58
UniProt ID: (Human) P43628
Entrez Gene ID: (Human) 3804
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support