Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ANT Antibodies

          Invitrogen

          ANT Polyclonal Antibody

          1 Reference
          View all (8) ANT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ANT Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ANT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ANT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ANT Antibody (PA5-102508) in IHC (P)

          Immunohistochemistry analysis of ANT in mouse kidney tissue. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with ANT polyclonal antibody (Product # PA5-102508) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ANT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANT Antibody in Western Blot (WB)
          ANT Polyclonal Antibody

          Product Details

          PA5-102508

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Bovine

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human SLC25A4(Accession P12235), corresponding to amino acid residues V78-L128.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2851910

          Product Specific Information

          Antibody detects endogenous levels of total SLC25A4.

          Target Information

          ANT1 catalyzes the exchange of ADP and ATP across the mitochondrial inner membrane.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; ADP ATP translocase; ADP,ATP carrier protein 1; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP.ATP translocase; ADP/ADT translocator protein; ADP/ATP carrier 1; ADP/ATP translocase 1; ANT 1; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; progressive external ophthalmoplegia 2; progressive external ophthalmoplegia 3; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_1; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; Solute carrier family 25 member 4; unnamed protein product

          View more View less

          Gene Aliases: AAC1; Anc1; ANT; ANT 1; ANT1; AU019225; MTDPS12; MTDPS12A; PEO2; PEO3; PEOA2; SLC25A4; T1

          View more View less

          UniProt ID: (Human) P12235, (Rat) Q05962, (Mouse) P48962

          View more View less

          Entrez Gene ID: (Human) 291, (Rat) 85333, (Mouse) 11739

          View more View less

          Function(s)
          ATP:ADP antiporter activity protein binding hydrogen ion transmembrane transporter activity adenine transmembrane transporter activity antiporter activity oxidative phosphorylation uncoupler activity transporter activity enzyme binding transfer/carrier protein
          Process(es)
          mitochondrial genome maintenance generation of precursor metabolites and energy apoptotic mitochondrial changes adenine transport ADP transport regulation of mitochondrial membrane permeability transmembrane transport negative regulation of necroptotic process mitochondrial ADP transmembrane transport negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway positive regulation of mitophagy hydrogen ion transmembrane transport mitochondrial ATP transmembrane transport adaptive thermogenesis liver development heart development aging negative regulation of cardiac muscle cell apoptotic process positive regulation of cell growth involved in cardiac muscle cell development response to interleukin-1 negative regulation of mitochondrial membrane permeability involved in apoptotic process positive regulation of oxidative phosphorylation uncoupler activity transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline