Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total LHX3.
This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp762A2013; Homeobox protein LIM-3; Homeobox protein P-LIM; LHX3a; LHX3b; LIM homeo box protein 3; LIM homeobox protein 3; LIM/homeobox protein Lhx3; LIM/homeodomain protein LHX3
Gene Aliases: CPHD3; LHX3; Lim-3; LIM3; M2-LHX3; mLim-3; mLIM3; P-LIM; Plim
UniProt ID: (Human) Q9UBR4, (Mouse) P50481
Entrez Gene ID: (Human) 8022, (Rat) 170671, (Mouse) 16871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support