Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total SOX8/9/17/18.
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mSox9; ragged; SOX18; SOX9; SRY (sex determining region Y)-box 17; SRY (sex determining region Y)-box 18; SRY (sex determining region Y)-box 8; SRY (sex determining region Y)-box 9; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY (sex determining region Y)-box9; SRY (sex-determining region Y)-box 9 protein; SRY-box 17; SRY-box 18; SRY-box 8; SRY-box 9; SRY-box containing gene 17; SRY-box containing gene 18; SRY-box containing gene 8; SRY-box containing gene 9; Sry-related HMG-box gene 18; SRY-related HMG-box transcription factor SOX17; SRY-related HMG-box, gene 9; Transcription factor SOX-17; Transcription factor SOX-18; Transcription factor SOX-8; Transcription factor SOX-9; unnamed protein product
Gene Aliases: 2010306G03Rik; AI385749; AV220920; CMD1; CMPD1; ENH13; HLTRS; HLTS; mKIAA4243; PPH7; Ra; Ragl; Sox-17; Sox-18; Sox-8; Sox-9; SOX17; SOX18; SOX8; SOX9; SRA1; SRXX2; SRXY10; TES; TESCO; VUR3
UniProt ID: (Human) Q9H6I2, (Human) P35713, (Human) P57073, (Human) P48436, (Mouse) Q61473, (Mouse) P43680, (Mouse) Q04886, (Rat) F1LYL9, (Mouse) Q04887
Entrez Gene ID: (Human) 64321, (Human) 54345, (Human) 30812, (Human) 6662, (Mouse) 20671, (Rat) 312936, (Rat) 311723, (Mouse) 20672, (Mouse) 20681, (Rat) 302993, (Rat) 140586, (Mouse) 20682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support