Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total ALS2CR12.
ALS2CR12 is a protein coding gene and gene ontology (GO) annotation includes cytoplasm; outer dense fiber; sperm fibrous sheath; sperm flagellum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALS2 chromosome region 12; ALS2CR12; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 12 protein; amyotrophic lateral sclerosis 2 chromosome region 12; amyotrophic lateral sclerosis 2 chromosome region candidate 12; Flagellum-associated coiled-coil domain-containing protein 1; unnamed protein product
Gene Aliases: ALS2CR12; FLACC1
UniProt ID: (Human) Q96Q35
Entrez Gene ID: (Human) 130540
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support