Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 11
Antibody detects endogenous levels of total HEATR1.
Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10359; HEAT repeat-containing protein 1; Protein BAP28; protein UTP10; U3 small nucleolar RNA-associated protein 10 homolog; unnamed protein product; UTP10, small subunit (SSU) processome component, homolog
Gene Aliases: AA517551; B130016L12Rik; BAP28; BC019693; HEATR1; RGD1564769; UTP10
UniProt ID: (Human) Q9H583
Entrez Gene ID: (Human) 55127, (Mouse) 217995, (Rat) 361262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support