Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NOD2 Antibodies

          Invitrogen

          NOD2 Polyclonal Antibody

          1 Reference
          View all (16) NOD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NOD2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (30)
          NOD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NOD2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 30

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NOD2 Antibody (PA5-104317) in ICC/IF

          Immunocytochemistry analysis of NOD2 in A549 cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with NOD2 polyclonal antibody (Product # PA5-104317) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG(H+L) (Red) and an AlexaFluor488 conjugated goat anti-mouse IgG(H+L) (Green) were used as the secondary antibodies. The nuclear counter stain is DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NOD2 Antibody in Immunocytochemistry (ICC/IF)
          NOD2 Antibody in Immunocytochemistry (ICC/IF)
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NOD2 Antibody in Western Blot (WB)
          NOD2 Antibody in Western Blot (WB)
          NOD2 Polyclonal Antibody

          Product Details

          PA5-104317

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human NOD2(Accession Q9HC29), corresponding to amino acid residues A110-G160.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2853627

          Product Specific Information

          Antibody detects endogenous levels of total NOD2.

          Target Information

          NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: caspase recruitment domain family, member 15; caspase recruitment domain protein 15; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; NOD 2; NOD-2; NOD-like receptor C2; NOD2 protein; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; Nucleotide-binding oligomerization domain-containing protein 2; unnamed protein product

          View more View less

          Gene Aliases: ACUG; BLAU; BLAUS; CARD15; CD; CLR16.3; F830032C23Rik; IBD1; NLRC2; NOD2; NOD2B; PSORAS1; YAOS

          View more View less

          UniProt ID: (Human) Q9HC29

          View more View less

          Entrez Gene ID: (Human) 64127, (Rat) 291912, (Mouse) 257632

          View more View less

          Function(s)
          nucleotide binding actin binding protein binding ATP binding enzyme binding protein kinase binding Hsp70 protein binding muramyl dipeptide binding pattern recognition receptor activity peptidoglycan binding ubiquitin binding anion binding ADP binding macromolecular complex binding CARD domain binding Hsp90 protein binding carbohydrate derivative binding endodeoxyribonuclease activity, producing 5'-phosphomonoesters scaffold/adaptor protein non-motor microtubule binding protein
          Process(es)
          temperature homeostasis pattern recognition receptor signaling pathway adaptive immune response immune system process positive regulation of dendritic cell antigen processing and presentation positive regulation of cytokine production involved in immune response positive regulation of dendritic cell cytokine production positive regulation of type 2 immune response autophagy defense response I-kappaB kinase/NF-kappaB signaling response to nutrient positive regulation of cell proliferation detection of biotic stimulus detection of bacterium maintenance of gastrointestinal epithelium regulation of appetite response to muramyl dipeptide detection of muramyl dipeptide positive regulation of interleukin-1 beta production positive regulation of interleukin-10 production positive regulation of interleukin-17 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production positive regulation of stress-activated MAPK cascade intracellular signal transduction intestinal stem cell homeostasis p38MAPK cascade defense response to bacterium regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of MAPK cascade innate immune response positive regulation of Notch signaling pathway positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade positive regulation of gamma-delta T cell activation host-mediated regulation of intestinal microbiota composition positive regulation of epithelial cell proliferation regulation of inflammatory response positive regulation of B cell activation positive regulation of NF-kappaB transcription factor activity positive regulation of ERK1 and ERK2 cascade nucleotide-binding oligomerization domain containing 2 signaling pathway protein K63-linked ubiquitination cellular response to lipopolysaccharide cellular response to peptidoglycan cellular response to muramyl dipeptide protein linear polyubiquitination antibacterial innate immune response positive regulation of cytokine production involved in inflammatory response positive regulation of NIK/NF-kappaB signaling positive regulation of mitophagy positive regulation of protein K63-linked ubiquitination negative regulation of macrophage apoptotic process innate immune response in mucosa activation of immune response cytokine production involved in immune response immunoglobulin production involved in immunoglobulin mediated immune response negative regulation of T cell mediated immunity negative regulation of inflammatory response to antigenic stimulus positive regulation of humoral immune response mediated by circulating immunoglobulin positive regulation of antibacterial peptide biosynthetic process positive regulation of biosynthetic process of antibacterial peptides active against Gram-positive bacteria positive regulation of protein ubiquitination negative regulation of NF-kappaB transcription factor activity response to peptidoglycan response to lipopolysaccharide negative regulation of interferon-gamma production negative regulation of interleukin-12 production negative regulation of interleukin-18 production negative regulation of interleukin-2 production negative regulation of tumor necrosis factor production positive regulation of interleukin-12 production negative regulation of toll-like receptor 2 signaling pathway activation of MAPK activity involved in innate immune response response to exogenous dsRNA positive regulation of phosphatidylinositol 3-kinase activity negative regulation of growth of symbiont in host positive regulation of innate immune response positive regulation of interleukin-1 beta secretion positive regulation of peptidyl-tyrosine phosphorylation positive regulation of phagocytosis defense response to Gram-positive bacterium positive regulation of oxidoreductase activity positive regulation of nitric-oxide synthase biosynthetic process positive regulation of prostaglandin-endoperoxide synthase activity cellular response to organic cyclic compound macrophage inflammatory protein-1 alpha production regulation of neutrophil chemotaxis positive regulation of prostaglandin-E synthase activity cytokine secretion involved in immune response inflammatory response signal transduction protein oligomerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline