Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total OSGI1.
OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMSC-derived growth inhibitor; Bone marrow stromal cell-derived growth inhibitor; huOKL38; HuOKL38-1a; HuOKL38-2a; HuOKL38-2b; Ovary, kidney and liver protein 38; Oxidative stress-induced growth inhibitor 1; pregnancy-induced growth inhibitor; Pregnancy-induced growth inhibitor OKL38; unnamed protein product
Gene Aliases: 1700012B18Rik; BDGI; OKL38; OSGIN1
UniProt ID: (Human) Q9UJX0, (Mouse) Q8VC10
Entrez Gene ID: (Human) 29948, (Mouse) 71839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support