Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UBE2A/UBE2B Antibodies

          Invitrogen

          UBE2A/UBE2B Polyclonal Antibody

          View all (3) UBE2A/UBE2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UBE2A/UBE2B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          UBE2A/UBE2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          UBE2A/UBE2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UBE2A/UBE2B Antibody (PA5-104442) in IHC (P)

          Immunohistochemistry analysis of UBE2A/UBE2B in rat testis tissue. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with UBE2A/UBE2B polyclonal antibody (Product # PA5-104442) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UBE2A/UBE2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBE2A/UBE2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBE2A/UBE2B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBE2A/UBE2B Antibody in Western Blot (WB)
          UBE2A/UBE2B Polyclonal Antibody

          Product Details

          PA5-104442

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human UBE2B(Accession P63146), corresponding to amino acid residues N94-I144.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816021

          Product Specific Information

          Antibody detects endogenous levels of total UBE2A/B.

          Target Information

          The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme A; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; HHR6A (Human homologue of yeast RAD 6); putative; HHR6B (Human homologue of yeast RAD 6); putative; HR6A; HR6B; RAD6 homolog A; RAD6 homolog B; UBE2B; Ubiquitin carrier protein A; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 A; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2A; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-conjugating enzyme HR6A; Ubiquitin-protein ligase A; Ubiquitin-protein ligase B; unnamed protein product

          View more View less

          Gene Aliases: 2610301N02Rik; E2-14k; E2-17kDa; HHR6A; HHR6B; HR6A; HR6B; Mhr6a; mHR6B; MRXS30; MRXSN; RAD6A; RAD6B; UBC2; UBCH1; UBE2A; UBE2B

          View more View less

          UniProt ID: (Human) P49459, (Human) P63146, (Mouse) Q9Z255, (Rat) P63149, (Mouse) P63147

          View more View less

          Entrez Gene ID: (Human) 7319, (Human) 7320, (Rat) 298317, (Mouse) 22209, (Rat) 81816, (Mouse) 22210

          View more View less

          Function(s)
          nucleotide binding ubiquitin-protein transferase activity protein binding ATP binding transferase activity ubiquitin-like protein transferase activity ubiquitin protein ligase binding ubiquitin binding ubiquitin conjugating enzyme activity molecular_function ligase activity ubiquitin protein ligase activity
          Process(es)
          protein polyubiquitination response to hypoxia in utero embryonic development DNA repair postreplication repair maintenance of chromatin silencing ubiquitin-dependent protein catabolic process protein monoubiquitination cellular response to DNA damage stimulus spermatogenesis sperm axoneme assembly response to UV positive regulation of reciprocal meiotic recombination protein ubiquitination histone ubiquitination regulation of histone modification cellular response to insulin stimulus negative regulation of histone phosphorylation histone H2A ubiquitination response to drug negative regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of cAMP-mediated signaling protein stabilization chiasma assembly protein autoubiquitination canonical Wnt signaling pathway histone lysine demethylation synaptonemal complex organization protein K63-linked ubiquitination protein K48-linked ubiquitination protein K11-linked ubiquitination biological_process positive regulation of cell proliferation maternal process involved in female pregnancy G2/M transition of mitotic cell cycle cytoplasmic translation chromatin organization chromatin remodeling positive regulation of mitophagy apoptotic process response to xenobiotic stimulus ectopic germ cell programmed cell death meiotic telomere clustering negative regulation of developmental process positive regulation of canonical Wnt signaling pathway negative regulation of post-translational protein modification negative regulation of reproductive process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline