Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • LIMK1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Phospho-LIMK1 (Thr508) Polyclonal Antibody

          View all (39) LIMK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support

          Cite Phospho-LIMK1 (Thr508) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (43)
          Phospho-LIMK1 (Thr508) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-LIMK1 (Thr508) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-LIMK1 (Thr508) Antibody (PA5-104925) in ICC/IF

          Immunofluorescent analysis of Phospho-LIMK1 (Thr508) in NIH-3T3. Samples were fixed with paraformaldehyde, permeabilized with 0.1% Triton X-100, blocked with 10% serum (45 min at 25°C) incubated with Phospho-LIMK1 (Thr508) polyclonal antibody (Product # PA5-104925) using a dilution of 1:200 (1 hr, 37°C), and followed by goat anti-rabbit IgG Alexa Fluor 594 at a dilution of 1:600. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-LIMK1 (Thr508) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-LIMK1 (Thr508) Antibody in Immunohistochemistry (IHC)
          Phospho-LIMK1 (Thr508) Antibody in Western Blot (WB)
          Phospho-LIMK1 (Thr508) Polyclonal Antibody

          Product Details

          PA5-104925

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human LIMK1 around the phosphorylation site of Thr508.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          RRID

          AB_2816398

          Target Information

          LIMK1 is a serine/threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK1 has also been implicated in brain development. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline