Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • BRK Antibodies

          Invitrogen

          Phospho-BRK (Tyr447) Polyclonal Antibody

          View all (19) BRK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-BRK (Tyr447) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (37)
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 37

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-BRK (Tyr447) Antibody (PA5-105207) in IHC (P)

          Immunohistochemistry analysis of Phospho-BRK (Tyr447) in mouse colorectal tissue. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with Phospho-BRK (Tyr447) polyclonal antibody (Product # PA5-105207) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-BRK (Tyr447) Antibody in Western Blot (WB)
          Phospho-BRK (Tyr447) Polyclonal Antibody

          Product Details

          PA5-105207

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human PTK6(Accession Q13882), corresponding to amino acid residues around phosphorylated Tyr447.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816680

          Product Specific Information

          Antibody detects endogenous levels of BRK only when phosphorylated at Tyr447.

          Target Information

          Breast tumor kinase (Brk) is a non-receptor protein tyrosine kinase expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines. Brk is 80% identity to Sik (Src-related intestinal kinase) and exhibits the features of a novel non-receptor tyrosine kinase, including amino terminal SH3 and SH2 domains. Low levels of Brk transcripts have been identified in some human breast tumor cell lines but not in normal breast tissue.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALT-PTK6; Breast tumor kinase; deltam5; FLJ42088; protein tyrosine kinase 6; Protein-tyrosine kinase 6; protein-tyrosine kinase BRK; PTK6 protein tyrosine kinase 6; SRC-related intestinal kinase; tyrosine kinase identified from skin; Tyrosine-protein kinase BRK; unnamed protein product

          View more View less

          Gene Aliases: BRK; PTK6; Sik; tks; Tksk

          View more View less

          UniProt ID: (Human) Q13882, (Mouse) Q64434

          View more View less

          Entrez Gene ID: (Human) 5753, (Mouse) 20459, (Rat) 366275

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding kinase activity transferase activity identical protein binding transferase activity, transferring phosphorus-containing groups
          Process(es)
          protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of neuron projection development phosphorylation cell migration actin cytoskeleton organization peptidyl-tyrosine autophosphorylation regulation of cell proliferation tyrosine phosphorylation of Stat3 protein tyrosine phosphorylation of Stat5 protein innate immune response negative regulation of growth protein autophosphorylation intestinal epithelial cell differentiation negative regulation of protein tyrosine kinase activity cellular response to retinoic acid tyrosine phosphorylation of STAT protein cell differentiation ERBB2 signaling pathway positive regulation of DNA replication positive regulation of epidermal growth factor receptor signaling pathway positive regulation of cell cycle positive regulation of JAK-STAT cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline