Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total WRAP53.
TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Telomerase Cajal body protein 1; unnamed protein product; WD repeat containing, antisense to TP53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; WRAP53beta
Gene Aliases: BC021790; DKCB3; TCAB1; WDR79; WRAP53
UniProt ID: (Human) Q9BUR4, (Rat) Q5XII5, (Mouse) Q8VC51
Entrez Gene ID: (Human) 55135, (Rat) 287432, (Mouse) 216853
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support