Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Antibody detects endogenous levels of total ZNF18.
ZNF18 be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDSG1; Heart development-specific gene 1 protein; unnamed protein product; Zinc finger protein 18; zinc finger protein 18 (KOX 11); Zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6
Gene Aliases: 1700128E15Rik; D11Ertd714e; HDSG1; KOX11; Zfp18; Zfp535; ZKSCAN6; ZNF18; ZNF535; ZSCAN38
UniProt ID: (Human) P17022, (Rat) Q642B9, (Mouse) Q810A1
Entrez Gene ID: (Human) 7566, (Rat) 303226, (Mouse) 52712
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support