Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KV1.5 (KCNA5) Antibodies

          Invitrogen

          KV1.5 (KCNA5) Polyclonal Antibody

          View all (18) KV1.5 (KCNA5) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite KV1.5 (KCNA5) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          KV1.5 (KCNA5) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KV1.5 (KCNA5) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KV1.5 (KCNA5) Antibody (PA5-109279) in ICC/IF

          Immunofluorescent analysis of KCNA5 in SKOV-3 cells (green). Samples were fixed in paraformaldehyde and permeabilised with 0.25% Triton X100/PBS, incubated with KCNA5 polyclonal antibody (Product # PA5-109279), followed by DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KV1.5 (KCNA5) Antibody in Immunocytochemistry (ICC/IF)
          KV1.5 (KCNA5) Antibody in Immunocytochemistry (ICC/IF)
          KV1.5 (KCNA5) Antibody in Immunocytochemistry (ICC/IF)
          KV1.5 (KCNA5) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KV1.5 (KCNA5) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KV1.5 (KCNA5) Antibody in Western Blot (WB)
          KV1.5 (KCNA5) Antibody in Flow Cytometry (Flow)
          KV1.5 (KCNA5) Polyclonal Antibody

          Product Details

          PA5-109279

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:800
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide within rat KCNA5 aa 249-298.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol, 0.2% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2854690

          Product Specific Information

          Positive Control: PANC-1, LOVO, SKOV-3, rat skeletal muscle tissue, rat heart tissue.

          Target Information

          In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cardiac potassium channel; HPCN1; insulinoma and islet potassium channel; MGC117058; MGC117059; Potassium (K+) channel protein alpha 5; potassium channel 1; potassium channel, voltage gated shaker related subfamily A, member 5; potassium voltage gated channel, shaker related subfamily, member 5; Potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; Voltage-gated potassium channel HK2; voltage-gated potassium channel protein Kv1.5; Voltage-gated potassium channel subunit Kv1.5

          View more View less

          Gene Aliases: ATFB7; HCK1; HK2; HPCN1; KCNA5; Kv1; KV1.5; PCN1

          View more View less

          UniProt ID: (Human) P22460, (Rat) P19024

          View more View less

          Entrez Gene ID: (Human) 3741, (Rat) 25470

          View more View less

          Function(s)
          receptor binding ion channel activity voltage-gated potassium channel activity delayed rectifier potassium channel activity potassium channel activity protein binding outward rectifier potassium channel activity protein kinase binding alpha-actinin binding voltage-gated potassium channel activity involved in bundle of His cell action potential repolarization voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization voltage-gated potassium channel activity involved in SA node cell action potential repolarization scaffold protein binding potassium channel inhibitor activity voltage-gated ion channel
          Process(es)
          action potential response to hypoxia ion transport potassium ion transport Notch signaling pathway response to mechanical stimulus regulation of vasoconstriction ion transmembrane transport regulation of membrane potential response to hydrogen peroxide regulation of insulin secretion protein oligomerization protein homooligomerization negative regulation of cytosolic calcium ion concentration potassium ion homeostasis transmembrane transport response to hyperoxia membrane hyperpolarization regulation of atrial cardiac muscle cell membrane repolarization potassium ion transmembrane transport atrial cardiac muscle cell action potential membrane repolarization during bundle of His cell action potential membrane repolarization during SA node cell action potential regulation of heart rate by cardiac conduction potassium ion export across plasma membrane membrane repolarization during atrial cardiac muscle cell action potential positive regulation of G1/S transition of mitotic cell cycle positive regulation of myoblast proliferation response to organic substance regulation of potassium ion transport negative regulation of potassium ion transport potassium ion export
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline