Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: VLPDFSTIKK GFCKPREEMV LSGKYKSGEQ ILRLANERFA VPEILFNPSD IGIQEMGIPE AIVYSIQNLP EEMQPHFFKN IVLTGGNSLF PGFRDRVYSE VRCLTPTDYD VSVVLPENPI TYAWEGGKLI SENDDFEDMV VTREDYEENG HSVCEEKFDI
The function of this protein remains unknown.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-related protein 6; ARP6 actin related protein 6 homolog; ARP6 actin-related protein 6 homolog; FLJ13433; h hARPX; hArp6; hARPX; mArp6; unnamed protein product
Gene Aliases: 2010200J04Rik; ACTR6; ARP6; ArpX; AU044443; CDA12; hARP6; hARPX; HSPC281; MSTP136
UniProt ID: (Human) Q9GZN1, (Mouse) Q9D864
Entrez Gene ID: (Human) 64431, (Rat) 314718, (Mouse) 67019
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support