Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MAFLKLRDQP SLVQAIFNGD PDEVRALIFK KEDVNFQDNE KRTPLHAAAY LGDAEIIELL ILSGARVNAK DSKWLTPLHR AVASCSEEAV QVLLKHSADV NARDKNWQTP LHIAAANKAV KCAEALVPLL SNVNVSDRAG RTALHHAAFS GHGEMVKLLL SRGANINAFD KKDRRAIHWA AYMGHIEVVK LLVSHGAEVT CKDKKSYTPL HAAASSGMIS VVKYLLDLGV DMNEPNAYGN TPLHVACYNG QDVVVNELID CGAIVNQKNE KGFTPLHFAA ASTHGALCLE LLVGNGADVN
Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. Involved in the PP6-mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha. Selectively inhibits the phosphatase activity of PPP1C. Targets PPP1C to modulate HNRPK phosphorylation. ANKRD28 selectively regulated phosphorylation of hnRNP-K at ser284, an effect that was enhanced in vivo by an S1013A/S1017A double mutant, suggesting that ANKRD28 targets PP1 in a regulated manner to dephosphorylate hnRNP-K selectively at ser284.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 28; phosphatase interactor targeting K protein; Phosphatase interactor targeting protein hnRNP K; PITK; PP6-ARS-A; protein phosphatase 1, regulatory subunit 65; protein phosphatase 6 ankyrin repeat subunit A; Serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A; serine/threonine-protein phosphatase 6 regulatory subunit ARS-A; unnamed protein product
Gene Aliases: AI465466; AI604979; ANKRD28; CFAP79; E430019N21Rik; FAP79; KIAA0379; mKIAA0379; PITK; PPP1R65
UniProt ID: (Human) O15084, (Mouse) Q505D1
Entrez Gene ID: (Human) 23243, (Mouse) 105522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support