Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Hydrolyzes glucose-6-phosphate to glucose in the endoplasmic reticulum. May form with the glucose-6-phosphate transporter (SLC37A4/G6PT) a ubiquitously expressed complex responsible for glucose production through glycogenolysis and gluconeogenesis. Probably required for normal neutrophil function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.1.3.9; G-6-Pase 3; G6Pase 3; G6Pase-beta; Glc-6-Pase-beta; glucose 6 phosphatase, catalytic, 3; Glucose-6-phosphatase 3; Glucose-6-phosphatase beta; glucose-6-phosphatase catalytic subunit 3; glucose-6-phosphatase catalytic subunit-related; glucose-6-phosphatase-beta; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein
Gene Aliases: 0710001K01Rik; AU019276; AU045429; AV128920; AW545836; G6PC3; SCN4; UGRP
UniProt ID: (Human) Q9BUM1, (Rat) Q6AZ83, (Mouse) Q6NSQ9
Entrez Gene ID: (Human) 92579, (Rat) 303565, (Mouse) 68401
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support