Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen sequence: DVVQEVQQVL SEFIRANPKA WAPVISAWSI DLMGQLSSTY SGQHQRVPHA TGALNELLQL WMGCRATRTL MDIYVQCLSA LIGSCPDACV DALLDTSVQH SPHFDWVVAH IGSSFPGTII SRVLSCGLKD FCVHGGAGGG AGSSGGSSSQ TPSTDPFPGS PAIPAEKRVP KIASVVGILG HLASRHGDSI RRELLRMFHD SLAGGSGGRS GDPSLQATVP FLLQLAVMSP ALLGTVSGEL VDCLKPPAVL SQLQQHLQGF PREELDNMLN LAVHLVSQAS G
INTS5 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Int5; Integrator complex subunit 5
Gene Aliases: 1110055N21Rik; INT5; INTS5; KIAA1698; mKIAA1698
UniProt ID: (Human) Q6P9B9, (Mouse) Q8CHT3
Entrez Gene ID: (Human) 80789, (Mouse) 109077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support