Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This gene encodes the heavy chain subunit of the pre-alpha-trypsin inhibitor complex. This complex may stabilize the extracellular matrix through its ability to bind hyaluronic acid. Polymorphisms of this gene may be associated with increased risk for schizophrenia and major depressive disorder. This gene is present in an inter-alpha-trypsin inhibitor family gene cluster on chromosome 3.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inter-alpha (globulin) inhibitor H3; inter-alpha (globulin) inhibitor, H3 polypeptide; inter-alpha-inhibitor H3 chain; inter-alpha-inhibitor heavy chain 3; Inter-alpha-trypsin inhibitor heavy chain H3; ITI heavy chain H3; ITI-HC3; pre-alpha (globulin) inhibitor, H3 polypeptide; pre-alpha-inhibitor heavy chain 3; pre-alpha-inhibitor, heavy chain 3; Serum-derived hyaluronan-associated protein; SHAP; unnamed protein product
Gene Aliases: AW108094; H3P; Intin3; ITI-HC3; Itih-3; ITIH3; PAIHC3; SHAP
UniProt ID: (Human) Q06033, (Mouse) Q61704, (Rat) Q63416
Entrez Gene ID: (Human) 3699, (Mouse) 16426, (Rat) 50693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support