Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MQYSHHCEHL LERLNKQREA GFLCDCTIVI GEFQFKAHRN VLASFSEYFG AIYRSTSENN VFLDQSQVKA DGFQKLLEFI YTGTLNLDSW NVKEIHQAAD YLKVEEVVTK CKIKMEDFAF IANPSSTEIS SITGNIELNQ QTCLLTLRDY NNREKSEVST DLIQANPKQG ALAKKSSQTK KKKKAFNSPK TGQNKTVQYP SDILENASVE LFLDANKLPT PVVEQVAQIN DNSELELTSV VENTFPAQDI VHTVTVKRKR GKSQPNCALK EHSMSNIASV KSPYEAENSG EELDQRYSKA
This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed in dendritic cells; Myoneurin; splice variant; unnamed protein product; Zinc finger and BTB domain-containing protein 31; zinc finger protein with BTB/POZ domain
Gene Aliases: 2810011C24Rik; AA415053; AI661031; AW049392; MYNN; Mynr; OSZF; SBBIZ1; ZBTB31; ZNF902
UniProt ID: (Human) Q9NPC7, (Mouse) Q99MD8
Entrez Gene ID: (Human) 55892, (Mouse) 80732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support