Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: SGAEIEDDDE EGFDDEDEFD DEHDDDLDTE DNELEELEER AEARKKTTEK QSAAALCVGV GSFADPDDLP GLAHFLEHMV FMGSLKYPDE NGFDAFLKKH GGSDNASTDC ERTVFQFDVQ RKYFKEALDR WAQFFIHPLM IRDAIDREVE AVDSEYQLAR PSDANRKEML FGSLARPGHP MGKFFWGNAE TLKHEPRKNN IDTHARLREF WMRYYSSHYM TLVVQSKETL DTLEKWVTEI F
Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-arginine dibasic convertase; N-arginine dibasic convertase 1; Nardilysin; nardilysin (N-arginine dibasic convertase); nardilysin 1 (N-arginine dibasic convertase); Nardilysin convertase; nardilysin, N-arginine dibasic convertase 1; nardilysin, N-arginine dibasic convertase, NRD convertase 1; NRD convertase; NRD-C; ORF1; unnamed protein product
Gene Aliases: 2600011I06Rik; AI875733; hNRD1; hNRD2; NRD-C; NRD1; NRDC
UniProt ID: (Human) O43847, (Mouse) Q8BHG1, (Rat) P47245
Entrez Gene ID: (Human) 4898, (Mouse) 230598, (Rat) 25499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support