Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MANENHGSPR EEASLLSHSP GTSNQSQPCS PKPIRLVQDL PEELVHAGWE KCWSRRENRP YYFNRFTNQS LWEMPVLGQH DVISDPLGLN ATPLPQDSSL VETPPAENKP RKRQLSEEQP SGNGVKKPKI EIPVTPTGQS VPSSPSIPGT PTLKMWGTSP EDKQQAALLR PTEVYWDLDI QTNAVIKHRG PSEVLPPHPE
PCIF1 may play a role in transcription elongation or in coupling transcription to pre-mRNA processing through its association with the phosphorylated C-terminal domain (CTD) of RNAPII largest subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cap-specific adenosine methyltransferase; CAPAM; hPCIF1; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; PDX-1 C terminus-interacting factor 1; PDX1 C-terminal inhibiting factor 1; phosphorylated CTD interacting factor PCIF1; Phosphorylated CTD-interacting factor 1; Protein phosphatase 1 regulatory subunit 121; protein phosphatase 1, regulatory subunit 121; unnamed protein product
Gene Aliases: 2310022K11Rik; C20orf67; CAPAM; F730014I05Rik; hCAPAM; hPCIF1; MT-A70; PCIF1; PPP1R121; RGD1310800
UniProt ID: (Human) Q9H4Z3, (Mouse) P59114
Entrez Gene ID: (Human) 63935, (Mouse) 228866, (Rat) 362269
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support