Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MSPVLHFYVR PSGHEGAAPG HTRRKLQGKL PELQGVETEL CYNVNWTAEA LPSAEETKKL MWLFGCPLLL DDVARESWLL PGSNDLLLEV GPRLNFSTPT STNIVSVCRA TGLGPVDRVE TTRRYRLSFA HPPSAEVEAI ALATLHDRMT EQHFPHPIQS FSPESMPEPL NGPINILGEG RLALEKANQE LGLALDSWDL DFYTKRFQEL QRNPSTVEAF DLAQSNSEHS RHWFFKGQLH VDGQKLVHSL FESIMSTQES SNPNNVLKFC DNSSAIQGKE VRFLRPEDPT RPSRFQQQQG
Purines are necessary for many cellular processes, including DNA replication, transcription, and energy metabolism. Ten enzymatic steps are required to synthesize inosine monophosphate (IMP) in the de novo pathway of purine biosynthesis. The enzyme encoded by this gene catalyzes the fourth step of IMP biosynthesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGAM synthase; FGAM synthetase; FGAR amidotransferase; Formylglycinamide ribonucleotide amidotransferase; Formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; IKZF1/PFAS fusion; KIAA0361; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase); Phosphoribosylformylglycineamide amidotransferase; pur4; unnamed protein product
Gene Aliases: 4432409B16Rik; FGAMS; FGAR-AT; FGARAT; GATD8; Gm18; KIAA0361; PFAS; PURL; Sofa
UniProt ID: (Human) O15067, (Mouse) Q5SUR0
Entrez Gene ID: (Human) 5198, (Rat) 287420, (Mouse) 237823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support