Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: HPVFDTKQIR NGVKSIGEKT EASLVWFTPP RTSNQWLDFW LRHRLQWWRK FAMSPSNFSS SDCQDEEGRK GNKLYYNFPW GKELIETLWN LGDHELLHMY PGNVSKLHGR DGRKNVVPCV LSVNGDLDRG MLAYLYDSFQ LTENSFTRKK NLHRKVLKLH PCLAPIKVAL DVGRGPTLEL RQVCQGLFNE LLENGISVWP GYLETMQSSL EQLYSKYDEM SILFTVLVTE TTLENGLIHL RSRDTTMKEM MHISKLKDFL IKYISSAKNV
This gene encodes the processivity subunit of the mitochondrial DNA polymerase gamma. The encoded protein forms a heterotrimer containing one catalytic subunit and two processivity subunits. This protein enhances DNA binding and promotes processive DNA synthesis. Mutations in this gene result in autosomal dominant progressive external ophthalmoplegia with mitochondrial DNA deletions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA polymerase gamma accessory 55 kDa subunit; DNA polymerase subunit gamma-2; DNA polymerase subunit gamma-2, mitochondrial; Hp55; Mitochondrial DNA polymerase accessory subunit; mitochondrial DNA polymerase subunit gamma-2; mitochondrial DNA polymerase, accessory subunit; mitochondrial polymerase gamma 2; MtPolB; p55; PolG-beta; polymerase (DNA directed), gamma 2, accessory subunit; polymerase (DNA) gamma 2, accessory subunit; polymerase, gamma 2; processivity factor for mitochondrial DNA polymerase catalytic subunit
Gene Aliases: HP55; MTDPS16; MTDPS16A; MTDPS16B; MTPOLB; PEOA4; POLB; POLG-BETA; POLG2; POLGB
UniProt ID: (Human) Q9UHN1, (Mouse) Q9QZM2
Entrez Gene ID: (Human) 11232, (Rat) 303612, (Mouse) 50776
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support