Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MRSPAVPTPA RGQLGVAFVL LPPHSEGARV FGALGPIGPS SPGLTLGGLA VSEHRLSNKL LAWSGVLEWQ EKRRPYSDST AKLKRTLPCQ AYVNQGENLE
PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator interaction domain-containing protein 2; Prostate tumor-overexpressed gene 1 protein; PTOV-1
Gene Aliases: ACID2; PP642; PTOV-1; PTOV1; UNQ6127/PRO20092
UniProt ID: (Human) Q86YD1
Entrez Gene ID: (Human) 53635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support