Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: QAAKQLSQNF LLDLRLTDKI VRKAGNLTNA YVYEVGPGPG GITRSILNAD VAELLVVEKD TRFIPGLQML SDAAPGKLRI VHGDVLTFKV EKAFSESLKR PWEDDPPNVH IIGNLPFSVS TPLIIKWLEN ISCRDGPFVY GRTQMTLTFQ KEVAERLAAN TGSKQRSRLS VMAQYLCNVR HIFTIPGQAF VPKPEVDVGV VHFTPLIQPK IEQPFKLVEK VVQNVFQFRR KYCHRGLRML FPEAQRLEST GRLLELADID PTLRPRQLSI SHFKSLCDVY RKMCDEDPQL FAYNFREELK RRKSKNEEKE EDDAENYRL
The protein encoded by this gene is a dimethyltransferase that methylates the conserved stem loop of mitochondrial 12S rRNA. The encoded protein also is part of the basal mitochondrial transcription complex and is necessary for mitochondrial gene expression. The methylation and transcriptional activities of this protein are independent of one another. Variations in this gene may influence the severity of aminoglycoside-induced deafness.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; Mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; mtTFB1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1; unnamed protein product
Gene Aliases: AI429207; CGI-75; CGI75; mtTFB; mtTFB1; TFB1M
UniProt ID: (Human) Q8WVM0, (Rat) Q811P6, (Mouse) Q8JZM0
Entrez Gene ID: (Human) 51106, (Rat) 308140, (Mouse) 224481
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support