Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Immunogen sequence: MAKDAGLIEA NGELKVFIDQ NLSPGKGVVS LVAVHPSTVN PLGKQLLPKT FGQSNVNIAQ QVVIGTPQRP AASNTLVVGS PHTPSTHFAS QNQPSDSSPW SAGKRNRKGE KNGKGLRHFS MKVCEKVQRK GTTSYNEVAD ELVAEFSAAD NHILPNESAY DQKNIRRRVY DALNVLMAMN IISKEKKEIK WIGLPTNSAQ ECQNLEVERQ RRLERIKQKQ SQLQELILQQ IAFKNLVQRN RHAEQQASRP PPPNSVIHLP FIIVNTSKKT VIDCSISNDK FEYLFNFDNT
DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AS1; ASRT1; cell cycle regulatory transcription factor DP1; down-regulated in liver cancer stem cells; DP; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Transcription factor Dp-1; unnamed protein product
Gene Aliases: DILC; Dp-1; DP1; DRTF1; TFDP1
UniProt ID: (Human) Q14186, (Mouse) Q08639
Entrez Gene ID: (Human) 7027, (Mouse) 21781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support