Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PCGAELTFEL PDNAKQCFHE EVEQGVKFSL DYQVITGGHY DVDCYVEDPQ GNTIYRETKK QYDSFTYRAE VKGVYQFCFS NEFSTFSHKT VYFDFQVGDE PPILPDMGNR VTALTQMESA CVTIHEALKT VIDSQTHYRL REAQDRARAE DLNSRVS
Potential role in vesicular protein trafficking, mainly in the early secretory pathway. Contributes to the coupled localization of TMED2 and TMED10 in the cis-Golgi network. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integral type I protein; Membrane protein p24B; MGC133022; p24 family protein gamma-4; P24B; p24gamma4; p26; testis tissue sperm-binding protein Li 48e; transmembrane emp24 domain containing 3; Transmembrane emp24 domain-containing protein 3; transmembrane emp24 protein transport domain containing 3; unnamed protein product
Gene Aliases: 1200002G13Rik; AW546672; C15orf22; P24B; p24g4; p24gamma4; p26; TMED3; UNQ5357/PRO1078
UniProt ID: (Human) Q9Y3Q3, (Mouse) Q78IS1, (Rat) Q6AY25
Entrez Gene ID: (Human) 23423, (Mouse) 66111, (Rat) 300888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support