Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: VCRVPGQIEV LRGFLEDVVP DATSAYPYLL LYESRQRRYL GSSPEGSGFC SKDRFVAYPC AVGQTAFSSG RHYWEVGMNI TGDALWALGV CRDNVSRKDR VPKCPENGFW VVQLSKGTKY LSTFSALTPV MLMEPPSHMG IFLDFEAGEV SFYSVSDGSH LHTYSQATFP GPLQPFFCLG APKSGQMVIS TVTMWVKG
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM17; RING finger protein 16; RING finger protein terf; RING finger-B box-coiled coil (RBCC) family member; RING-type E3 ubiquitin transferase TRIM17; Testis RING finger protein; tripartite motif protein 17; tripartite motif protein TRIM17; Tripartite motif-containing protein 17; unnamed protein product
Gene Aliases: RBCC; RNF16; TERF; TRIM17
UniProt ID: (Human) Q9Y577, (Rat) Q9WV59, (Mouse) Q7TPM3
Entrez Gene ID: (Human) 51127, (Rat) 64702, (Mouse) 56631
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support