Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH2 domain-containing adapter protein E; Src homology 2 domain containing E; src homology 2 domain-containing transforming protein E
Gene Aliases: 9430022A14; RGD1563897; SHE
UniProt ID: (Human) Q5VZ18, (Mouse) Q8BSD5
Entrez Gene ID: (Human) 126669, (Mouse) 214547, (Rat) 685088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support