Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The zinc transporter ZIP4, also known as SLC39A4, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP4 was initially identified as zinc-uptake transporter in the intestine and as a candidate gene for acrodermatitis enteropathica (AE), a rare inherited defect in the absorption of dietary zinc. Similar to other ZIP family members, aberrant expression of ZIP4 has been linked to cancer, specifically the progression and aggressiveness of pancreatic cancer. ZIP4 expression has also been shown to be activated in hepatocellular carcinomas, acting to repress apoptosis, enhance cell cycle and increase migration, suggesting ZIP4 may play a role in multiple types of cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activated in W/Wv mouse stomach 2; mAWMS2; solute carrier family 39 (zinc transporter) member 4 isoform SLC39A4_1; solute carrier family 39 (zinc transporter) member 4 isoform SLC39A4_2; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; unnamed protein product; zinc transporter 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4; ZRT/IRT-like protein 4
Gene Aliases: 1600025H15Rik; AEZ; AU041686; AWMS2; SLC39A4; ZIP4
UniProt ID: (Human) Q6P5W5, (Mouse) Q78IQ7
Entrez Gene ID: (Human) 55630, (Mouse) 72027
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support