Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mammalian INDY homolog; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaCT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; Sodium-coupled citrate transporter; Sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; Solute carrier family 13 member 5; unnamed protein product
Gene Aliases: DEE25; EIEE25; INDY; mINDY; NaC2/NaCT; NACT; SLC13A5
UniProt ID: (Human) Q86YT5, (Rat) Q8CJ44, (Mouse) Q67BT3
Entrez Gene ID: (Human) 284111, (Rat) 266998, (Mouse) 237831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support