Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Neuropeptide Y Receptor Y6 (NPY6R) is a receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibits adenylate cyclase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuropeptide Y receptor type 6; NPY6-R; Pancreatic polypeptide receptor 2; PP2; unnamed protein product
Gene Aliases: NPY1RL; Npy5r; Npy6r; NPY6RP; PP2; Ppyr2; Y2B
UniProt ID: (Mouse) Q61212
Entrez Gene ID: (Human) 4888, (Mouse) 18169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support